Learn More
Bioworld SpotQC hybridization Buffer, 20 mL
Catalog No. 50196839
Encompass_Preferred
SpotQC Hybridization Buffer is an optimized hybridization solution specifically formulated for use with IDT’s SpotQC detector oligos. It also allows for the dilution of the SpotQC detector oligo up to 20-fold while maintaining the superior detection of a broad range of oligo-probe lengths and GC compositions.
Catalog No. 50-196-839 Supplier Bioworld Supplier No. 107500171
SpotQC Hybridization Buffer is an optimized hybridization solution specifically formulated for use with IDT’s SpotQC detector oligos. It also allows for the dilution of the SpotQC detector oligo up to 20-fold while maintaining the superior detection of a broad range of oligo-probe lengths and GC compositions.