Learn More
Bioworld SpotQC hybridization Buffer, 125 mL
Catalog No. 50196840
Encompass_Preferred
SpotQC Hybridization Buffer is an optimized hybridization solution specifically formulated for use with IDT’s SpotQC detector oligos. It also allows for the dilution of the SpotQC detector oligo up to 20-fold while maintaining the superior detection of a broad range of oligo-probe lengths and GC compositions.
Catalog No. 50-196-840 Supplier Bioworld Supplier No. 107500172
SpotQC Hybridization Buffer is an optimized hybridization solution specifically formulated for use with IDT’s SpotQC detector oligos. It also allows for the dilution of the SpotQC detector oligo up to 20-fold while maintaining the superior detection of a broad range of oligo-probe lengths and GC compositions.