Learn More
Grainger Spotting Drill Bit Carbide 1/2
Catalog No. 502766019
Encompass_Preferred
The 16R641 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", from Grainger. It features an open web construction that cleans effectively without scratching. The pad is made of high-quality fibers and resin that resist shredding and matting. It can be used for light scrubbing and cleaning of surfaces. This durable hand pad measures 6 inches by 9 inches and has a maximum use temperature of 220°F. It effectively cleans a variety of surfaces including stainless steel, aluminum, ceramic, plastic and more. The 3M Scotch-Brite General Purpose Hand Pad is ideal for general cleaning in industrial, food service, and commercial environments.
Catalog No. 50-276-6019 Supplier Grainger Supplier No. 16R641
The 16R641 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", from Grainger. It features an open web construction that cleans effectively without scratching. The pad is made of high-quality fibers and resin that resist shredding and matting. It can be used for light scrubbing and cleaning of surfaces. This durable hand pad measures 6 inches by 9 inches and has a maximum use temperature of 220°F. It effectively cleans a variety of surfaces including stainless steel, aluminum, ceramic, plastic and more. The 3M Scotch-Brite General Purpose Hand Pad is ideal for general cleaning in industrial, food service, and commercial environments.