Learn More
Grainger Spotting Drill Bit Carbide 1/2
Catalog No. 502767464
Encompass_Preferred
The Grainger Approved Spotting Drill Bit is a high-speed steel spotting drill bit with a 1/8" shank diameter, 1/8" drill point diameter, and an overall length of 1-1/2". It has a straight shank and is designed for use on metal, wood, and plastics. The 135 degree split point tip helps prevent walking when starting a hole. This USA-made drill bit is ideal for precision hole location and provides accurate starts in automated equipment. It meets ANSI B94.11 standards.
Catalog No. 50-276-7464 Supplier Grainger Supplier No. 16R642
The Grainger Approved Spotting Drill Bit is a high-speed steel spotting drill bit with a 1/8" shank diameter, 1/8" drill point diameter, and an overall length of 1-1/2". It has a straight shank and is designed for use on metal, wood, and plastics. The 135 degree split point tip helps prevent walking when starting a hole. This USA-made drill bit is ideal for precision hole location and provides accurate starts in automated equipment. It meets ANSI B94.11 standards.