Learn More
Grainger Spotting Drill Bit Carbide 1/4
Supplier:  Grainger 16R625
Encompass_Preferred
The Micro 100 Spotting Drill Bit E-Drill is a high-speed steel spotting drill bit with a 1/8" shank diameter and a 60 degree included angle point. It has an overall length of 1-1/2" and is designed for center drilling and spotting operations in steel, cast iron, aluminum, brass, bronze and plastic. The E-Drill geometry allows for faster feed rates and has a 135 degree split point for easy centering and reduced thrust. This USA-made spotting drill bit can be run at speeds up to 39,000 RPM and is ideal for use on CNC machines, automatic screw machines, and manual machine tools.
Catalog No. 50-276-4981