Learn More
Grainger Spotting Drill Bit Carbide 1/4
Catalog No. 502765131
Encompass_Preferred
The Grainger Approved Spotting Drill Bit is a high-speed steel spotting drill bit with a 1/8" shank diameter and a 60 degree point angle. It has an overall length of 2-1/2" and is designed for use on steel, cast iron, aluminum, copper and brass. This USA-made spotting drill bit features a TiN coating for increased tool life and lubricity. It can be used for centering, spotting and scribing on manual and CNC machine tools. The Grainger item number for this spotting drill bit is 16R626.
Catalog No. 50-276-5131 Supplier Grainger Supplier No. 16R626
The Grainger Approved Spotting Drill Bit is a high-speed steel spotting drill bit with a 1/8" shank diameter and a 60 degree point angle. It has an overall length of 2-1/2" and is designed for use on steel, cast iron, aluminum, copper and brass. This USA-made spotting drill bit features a TiN coating for increased tool life and lubricity. It can be used for centering, spotting and scribing on manual and CNC machine tools. The Grainger item number for this spotting drill bit is 16R626.