Learn More
Grainger Spotting Drill Bit Carbide 3/4
Supplier:  Grainger 16R648
Encompass_Preferred
The Grainger Approved Spotting Drill Bit is a carbide tipped drill bit designed for spotting holes prior to drilling in metal. It has a 118 degree split point tip to help prevent walking and has an overall length of 2-1/2". The shank diameter is 1/4" and the drill diameter is #7. This is a high speed steel bit with cobalt added for increased hardness. It can drill through materials like steel, iron, aluminum and more. The Grainger item number for this product is 16R648. This spotting drill bit would be useful for creating pilot holes when drilling metal to increase accuracy and prevent walking.
Catalog No. 50-276-5760