Learn More
TARGETMOL CHEMICALS INC SPP-86 10MG
SDP
Catalog No. 502878812
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. SPP-86 is an effective and selective cell-permeable inhibitor of RET tyrosine kinase with an IC50 of 8 nM. Purity 98.19%
Catalog No. 50-287-8812 Supplier TARGETMOL CHEMICALS INC Supplier No. T1692310MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. SPP-86 is an effective and selective cell-permeable inhibitor of RET tyrosine kinase with an IC50 of 8 nM. Purity 98.19%