Learn More
Medchemexpress LLC HY-110193 5mg Medchemexpress, SPP-86 CAS:1357349-91-7 Purity:>98%
SDP
Catalog No. 501932347
Encompass_Preferred
Medchemexpress, HY-110193 5mg SPP-86 CAS:1357349-91-7 SPP-86 is a potent and selective cell permeable inhibitor of RET tyrosine kinase , with an IC 50 of 8 nM. SPP-86 inhibits RET-induced phosphatidylinositide 3-kinases (PI3K)/Akt and MAPK signaling, also inhibits RET-induced estrogen receptorα (ERα) phosphorylation in MCF7 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2347 Supplier Medchemexpress LLC Supplier No. HY1101935MG
Medchemexpress, HY-110193 5mg SPP-86 CAS:1357349-91-7 SPP-86 is a potent and selective cell permeable inhibitor of RET tyrosine kinase , with an IC 50 of 8 nM. SPP-86 inhibits RET-induced phosphatidylinositide 3-kinases (PI3K)/Akt and MAPK signaling, also inhibits RET-induced estrogen receptorα (ERα) phosphorylation in MCF7 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.