Learn More
Medchemexpress LLC SPP-DM1 | 452072-20-7 | 96.3% | 967.54 | C44H59ClN4O14S2 | 10 MG
SDP
Catalog No. 5000202745
Encompass_Preferred
SPP-DM1 is an antibody-drug conjugate (ADC) agent-linker conjugate that links the cytotoxic payload DM1 via an SPP linker to enable targeted delivery of a microtubule-disrupting agent. It is intended for research use in ADC development and preclinical antitumor studies, and is provided with analytical documentation to support characterization and handling.
  • Enables targeted delivery of a microtubule-disrupting payload.
  • Suitable for ADC development and preclinical antitumor research.
  • Provided with analytical documentation such as COA, HNMR, MS, and RP-HPLC.
  • Offered at defined purity suitable for research applications.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-02745 Supplier Medchemexpress LLC Supplier No. HY12649110MG
SPP-DM1 is an antibody-drug conjugate (ADC) agent-linker conjugate that links the cytotoxic payload DM1 via an SPP linker to enable targeted delivery of a microtubule-disrupting agent. It is intended for research use in ADC development and preclinical antitumor studies, and is provided with analytical documentation to support characterization and handling.
  • Enables targeted delivery of a microtubule-disrupting payload.
  • Suitable for ADC development and preclinical antitumor research.
  • Provided with analytical documentation such as COA, HNMR, MS, and RP-HPLC.
  • Offered at defined purity suitable for research applications.
  • Available in small pack sizes for laboratory use.