Learn More
Aviva Systems Biology Corp SPP1 Antibody - C-terminal region (ARP36677_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36677_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SPP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SPP1. Alternate names: OPN, BNSP, BSPI, ETA-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rat, Sheep
Catalog No. 50-014-56246