Learn More
ABclonal Technology SPPL2A Rabbit pAb
SDP
Supplier:  ABclonal Technology A155391000UL
Encompass_Preferred
This gene encodes a member of the GXGD family of aspartic proteases, which are transmembrane proteins with two conserved catalytic motifs localized within the membrane-spanning regions, as well as a member of the signal peptide peptidase-like protease (SPPL) family. This protein is expressed in all major adult human tissues and localizes to late endosomal compartments and lysosomal membranes. A pseudogene of this gene also lies on chromosome 15.
Catalog No. 50-001-06003