Learn More
Bachem Fmoc-Cys((R)-2,3-di(palmitoyloxy)-propyl)-OH
SDP
Catalog No. 501947146
Encompass_Preferred
Fmoc-Cys((R)-2,3-di(palmitoyloxy)-propyl)-OH, 139573-77-6, C53H83NO8S, Mr 894.31, 1g. Fmoc-(R)Pam2Cys-OH. SPPS employing the pure stereoisomer of Fmoc-Pam2Cys-OH allows to obtain more homogeneous lipopeptides. The configuration of the bis-palmitoyloxypropyl moiety could influence the biological activity of the peptide conjugate.
Catalog No. 50-194-7146 Supplier Bachem Supplier No. 4042951.1000
Fmoc-Cys((R)-2,3-di(palmitoyloxy)-propyl)-OH, 139573-77-6, C53H83NO8S, Mr 894.31, 1g. Fmoc-(R)Pam2Cys-OH. SPPS employing the pure stereoisomer of Fmoc-Pam2Cys-OH allows to obtain more homogeneous lipopeptides. The configuration of the bis-palmitoyloxypropyl moiety could influence the biological activity of the peptide conjugate.