Learn More
Medchemexpress LLC SPR206 acetate | 2408422-41-1 | 98.9% | 1 MG
SDP
Catalog No. 5000340523
Encompass_Preferred
SPR206 acetate is a polymyxin analog exhibiting antibiotic activity against Gram-negative pathogens, including multidrug-resistant variants. It works by interacting with the bacterium's outer membrane, demonstrating efficacy against a range of bacteria.
  • Antibiotic activity against Gram-negative pathogens
  • Effective against multidrug-resistant (MDR) variants
  • Interacts with bacterium's outer membrane
  • Solid, white to off-white appearance
  • Soluble in DMSO (≥ 100 mg/mL)
  • Shown to reduce bacterial burden in in vivo models
Catalog No. 50-003-40523 Supplier Medchemexpress LLC Supplier No. HY128780B1MG
SPR206 acetate is a polymyxin analog exhibiting antibiotic activity against Gram-negative pathogens, including multidrug-resistant variants. It works by interacting with the bacterium's outer membrane, demonstrating efficacy against a range of bacteria.
  • Antibiotic activity against Gram-negative pathogens
  • Effective against multidrug-resistant (MDR) variants
  • Interacts with bacterium's outer membrane
  • Solid, white to off-white appearance
  • Soluble in DMSO (≥ 100 mg/mL)
  • Shown to reduce bacterial burden in in vivo models