Learn More
Medchemexpress LLC SPR719 | 1384984-18-2 | 99.4% | 10 MG
SDP
Catalog No. 5000339423
Encompass_Preferred
SPR719 (VXc-486) is an orally active gyrase B inhibitor with bactericidal activity. It potently inhibits multiple agent-sensitive and drug-resistant isolates of Mycobacterium tuberculosis, making it promising for research into lung diseases caused by non-tuberculous mycobacteria (NTM).
  • Potently inhibits multiple agent-sensitive and drug-resistant isolates of Mycobacterium tuberculosis.
  • Shows bactericidal activity.
  • Research compound for lung diseases caused by non-tuberculous mycobacteria (NTM).
  • Orally active gyrase B inhibitor.
  • Inhibits M. tuberculosis replication in macrophages.
Catalog No. 50-003-39423 Supplier Medchemexpress LLC Supplier No. HY1293010MG
SPR719 (VXc-486) is an orally active gyrase B inhibitor with bactericidal activity. It potently inhibits multiple agent-sensitive and drug-resistant isolates of Mycobacterium tuberculosis, making it promising for research into lung diseases caused by non-tuberculous mycobacteria (NTM).
  • Potently inhibits multiple agent-sensitive and drug-resistant isolates of Mycobacterium tuberculosis.
  • Shows bactericidal activity.
  • Research compound for lung diseases caused by non-tuberculous mycobacteria (NTM).
  • Orally active gyrase B inhibitor.
  • Inhibits M. tuberculosis replication in macrophages.