Learn More
Medchemexpress LLC HY-12930 5mg Medchemexpress, SPR719 CAS:1384984-18-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129305MG
Encompass_Preferred
Medchemexpress, HY-12930 5mg SPR719 CAS:1384984-18-2 SPR719 (VXc-486) is a gyrase B inhibitor, with bactericidal activity. SPR719 potently inhibits multiple drug-sensitive isolates and drug-resistant isolates of Mycobacterium tuberculosis, with MICs of 0.03 to 0.30 μg/ml and 0.08 to 5.48 μg/ml, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1201