Learn More
Medchemexpress LLC SPR719 (VXc-486) | 1384984-18-2 | 99.4% | 100 MG
SDP
Catalog No. 5000339325
Encompass_Preferred
SPR719 (VXc-486) is an orally active gyrase B inhibitor with bactericidal activity. It shows promise for research into lung disease caused by non-tuberculous mycobacteria (NTM). It potently inhibits multiple agent-sensitive and drug-resistant isolates of *Mycobacterium tuberculosis*.
  • Potently inhibits *Mycobacterium tuberculosis*
  • Active against agent-sensitive and drug-resistant isolates
  • Promising for research into lung disease caused by non-tuberculous mycobacteria
  • Inhibits *M. tuberculosis* replication in macrophages
Catalog No. 50-003-39325 Supplier Medchemexpress LLC Supplier No. HY12930100MG
SPR719 (VXc-486) is an orally active gyrase B inhibitor with bactericidal activity. It shows promise for research into lung disease caused by non-tuberculous mycobacteria (NTM). It potently inhibits multiple agent-sensitive and drug-resistant isolates of *Mycobacterium tuberculosis*.
  • Potently inhibits *Mycobacterium tuberculosis*
  • Active against agent-sensitive and drug-resistant isolates
  • Promising for research into lung disease caused by non-tuberculous mycobacteria
  • Inhibits *M. tuberculosis* replication in macrophages