Learn More
Medchemexpress LLC HY-P1649B 5mg Medchemexpress, SPR741 (acetate) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1649B5MG
Encompass_Preferred
Medchemexpress, HY-P1649B 5mg SPR741 (acetate) CAS:0 SPR741 acetate (NAB741 acetate) is a cationic peptide derived from polymyxin B and is a potentiator molecule. SPR741 acetate increases the permeability of the outer membrane of Gram-negative bacteria and is used to treat severe Gram-negative bacteria infections. SPR741 acetate inhibits multidrug-resistant Gram-negative bacteria. The spectrum of activity of the antibiotic can be widened when used in combination with SPR741 acetate[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1695