Learn More
Medchemexpress LLC SPR741 acetate (NAB741 acetate) | 99.4% | 1052.18 | 50 MG
SDP
Catalog No. 5000423521
Encompass_Preferred
SPR741 acetate is a cationic peptide derived from polymyxin B, functioning as a potentiator molecule. It increases the permeability of the outer membrane of Gram-negative bacteria, making it suitable for treating severe Gram-negative bacterial infections.
  • Derived from polymyxin B
  • Functions as a potentiator molecule
  • Increases outer membrane permeability of Gram-negative bacteria
  • Used for treating severe Gram-negative bacterial infections
  • Inhibits multidrug-resistant Gram-negative bacteria
  • Combination with other antibiotics broadens their spectrum of activity
Catalog No. 50-004-23521 Supplier Medchemexpress LLC Supplier No. HYP1649B50MG
SPR741 acetate is a cationic peptide derived from polymyxin B, functioning as a potentiator molecule. It increases the permeability of the outer membrane of Gram-negative bacteria, making it suitable for treating severe Gram-negative bacterial infections.
  • Derived from polymyxin B
  • Functions as a potentiator molecule
  • Increases outer membrane permeability of Gram-negative bacteria
  • Used for treating severe Gram-negative bacterial infections
  • Inhibits multidrug-resistant Gram-negative bacteria
  • Combination with other antibiotics broadens their spectrum of activity