Learn More
Cayman Chemical (S)-Pramipexole (hydrochloride), Purity: ≥98%, CAS: 104632-25-9, 50 mg
SDP
Catalog No. 502955754
Encompass_Preferred
A dopamine D2S, D2L, D3, and D4 receptor agonist (EC50s = 426.58, 338.84, 2.24, and 128.82 nM, respectively, in a [35S]GTPγS binding assay); a partial agonist of α2A-ARs (EC50 = 3,548.13 nM); selective for dopamine D2-4 receptors (Kis = 954.99, 1,698.24, 12.59, 128.82 nM for for D2S, D2L, D3, and D4 receptors, respectively, in a radioligand binding assay) over D1 and D5 receptors (Kis = >10,000 nM for both); prevents MPTP-induced decreases in the number of dopaminergic neurons in the substantia nigra pars compacta in common marmosets at 60 μg/kg per day before, during, and after administration of MPTP
Catalog No. 50-295-5754 Supplier Cayman Chemical Supplier No. 1198150 MG
A dopamine D2S, D2L, D3, and D4 receptor agonist (EC50s = 426.58, 338.84, 2.24, and 128.82 nM, respectively, in a [35S]GTPγS binding assay); a partial agonist of α2A-ARs (EC50 = 3,548.13 nM); selective for dopamine D2-4 receptors (Kis = 954.99, 1,698.24, 12.59, 128.82 nM for for D2S, D2L, D3, and D4 receptors, respectively, in a radioligand binding assay) over D1 and D5 receptors (Kis = >10,000 nM for both); prevents MPTP-induced decreases in the number of dopaminergic neurons in the substantia nigra pars compacta in common marmosets at 60 μg/kg per day before, during, and after administration of MPTP