Learn More
Grainger Spray Adhesive 8 fl oz Bottle
Catalog No. 502763741
Encompass_Preferred
The ECLECTIC E6000 Spray Adhesive is a versatile, permanent spray adhesive that forms a strong bond on a variety of surfaces including fabric, wood, metal, glass, foam, and more. It applies easily with a controllable fine mist spray and sets quickly with no dripping. This adhesive is resistant to water and most solvents and remains flexible after drying. The 13 ounce aerosol can adheres approximately 15 square feet of material. E6000 is made in the USA and is a great multi-purpose adhesive for both indoor and outdoor applications.
Catalog No. 50-276-3741 Supplier Grainger Supplier No. 16NG97
The ECLECTIC E6000 Spray Adhesive is a versatile, permanent spray adhesive that forms a strong bond on a variety of surfaces including fabric, wood, metal, glass, foam, and more. It applies easily with a controllable fine mist spray and sets quickly with no dripping. This adhesive is resistant to water and most solvents and remains flexible after drying. The 13 ounce aerosol can adheres approximately 15 square feet of material. E6000 is made in the USA and is a great multi-purpose adhesive for both indoor and outdoor applications.