Learn More
Chemglass Life Sciences Spray Ball, 35mm, Fluid Driven, 3/8" Compression Fitting, PTFE
SDP
Catalog No. 501539701
Encompass_Preferred
This fluid driven Spray Ball gives nearly 360¿ spray coverage and is a convenient way to clean Chemglass 10 thru 100L glass reactors without dismantling the entire setup. It can also be used as a wall wetter. Spray ball is constructed of PTFE, the removable PTFE clip allows the rotating spray head to be easily removed for inspection and maintenance. 3/8¿ Top compression fitting is for connection to 3/8¿ FEP or PTFE tubing.The minimum joint clearance for 22mm and 36mm spray ball is 45/50.Please Note: Connecting Adapters And 3/8¿ PTFE Tubing Are Sold Separately.Features:Spray Ball/Head- Has Larger Slots for Improved Spray Pattern and Velocity of Spray. Larger Bore for More Flow. 3/8" Compression Fitting. All PTFE Body and Spray Ball. Pressure and Flow Information:CG-1989-X-35: 35mm Spray Ball -max tank diameter: 4.0ft @ 20 PSI (min pressure to spin): 10.45 L/min @ 50 PSI: 19.23 L/min
Catalog No. 50-153-9701 Supplier Chemglass Life Sciences Supplier No. CG1989X35
This fluid driven Spray Ball gives nearly 360¿ spray coverage and is a convenient way to clean Chemglass 10 thru 100L glass reactors without dismantling the entire setup. It can also be used as a wall wetter. Spray ball is constructed of PTFE, the removable PTFE clip allows the rotating spray head to be easily removed for inspection and maintenance. 3/8¿ Top compression fitting is for connection to 3/8¿ FEP or PTFE tubing.The minimum joint clearance for 22mm and 36mm spray ball is 45/50.Please Note: Connecting Adapters And 3/8¿ PTFE Tubing Are Sold Separately.Features:Spray Ball/Head- Has Larger Slots for Improved Spray Pattern and Velocity of Spray. Larger Bore for More Flow. 3/8" Compression Fitting. All PTFE Body and Spray Ball. Pressure and Flow Information:CG-1989-X-35: 35mm Spray Ball -max tank diameter: 4.0ft @ 20 PSI (min pressure to spin): 10.45 L/min @ 50 PSI: 19.23 L/min