Learn More
Ampco Spray Booth Scraper
SDP
Catalog No. 19174276 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
9 -1/4 in.
Catalog No. Length (English)
19-174-276 9 -1/4 in.
1 options
Catalog No. 19-174-276 Supplier Ampco Supplier No. S40
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant tool specially designed to remove dried paint, overspray, and debris from spray booths.
  • Made in the USA.
  • Width of blade tip: 3 in.
  • Overall length: 9 and 1/4 in.
Specifications
Product Type Scraper
Length (English) 9 -1/4 in.
Quantity 1 Ea.
Width (English) 3 in.