Learn More
BURKLE INC Spray bottle with hand pump, 8.45 oz. (250 ml), stroke: 0.04 oz. (1,2 ml)
Catalog No. 5000025746
Encompass_Preferred
High-quality, reusable hand spray bottle with rotating spray head, long spray lever and infinitely adjustable nozzle for fine mist or spot spray.
Ideal for cleaning, gardening, laboratories and more. Chemical resistant, with filter strainer and durable hand pump.
The hand spray bottle's wetted parts are made of PP, PE and stainless steel to ensure safe and reliable use. The spray bottle is highly resistant to a wide range of aggressive chemicals, both alkaline and acidic liquids.
Catalog No. 50-000-25746 Supplier BURKLE INC Supplier No. 03091025
High-quality, reusable hand spray bottle with rotating spray head, long spray lever and infinitely adjustable nozzle for fine mist or spot spray.
Ideal for cleaning, gardening, laboratories and more. Chemical resistant, with filter strainer and durable hand pump.
The hand spray bottle's wetted parts are made of PP, PE and stainless steel to ensure safe and reliable use. The spray bottle is highly resistant to a wide range of aggressive chemicals, both alkaline and acidic liquids.