Learn More
BURKLE INC Spray bottle Turn'n'Spray, 8.45 oz. (250 ml), stroke: 0.04 oz. (1,2 ml)
Catalog No. 5000025810
Encompass_Preferred
Thanks to its special technology, the Turn'n'Spray overhead spray bottle allows continuous spraying even overhead or at an angle.
Conventional sprayers cannot spray at an angle or overhead; as soon as the suction tube is out of the liquid, they stop spraying. Not so with our Turn'n'Spray overhead sprayer. It allows you to spray practically and conveniently from all directions, reaching even hard-to-reach places easily and without contortions. The spray bottle allows for comfortable and time-saving work.
The materials in contact with the fluid - PP, PE, silicone and stainless steel - are of high quality and guarantee durability, versatility and safe and reliable use. The spray bottle is resistant to a wide range of aggressive chemicals, both alkaline and acidic liquids.
Catalog No. 50-000-25810 Supplier BURKLE INC Supplier No. 03090002
Thanks to its special technology, the Turn'n'Spray overhead spray bottle allows continuous spraying even overhead or at an angle.
Conventional sprayers cannot spray at an angle or overhead; as soon as the suction tube is out of the liquid, they stop spraying. Not so with our Turn'n'Spray overhead sprayer. It allows you to spray practically and conveniently from all directions, reaching even hard-to-reach places easily and without contortions. The spray bottle allows for comfortable and time-saving work.
The materials in contact with the fluid - PP, PE, silicone and stainless steel - are of high quality and guarantee durability, versatility and safe and reliable use. The spray bottle is resistant to a wide range of aggressive chemicals, both alkaline and acidic liquids.