Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Spray Bottle Holder
SDP

Catalog No. 22000391
Change view
Click to view available options
Mounting Type:
Powerful Magnet
Powerful VHB Tape
Wall, Keyhole Slots
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-391 Powerful Magnet
22-000-392 Powerful VHB Tape
22-000-393 Wall, Keyhole Slots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-391 Supplier Poltex Supplier No. SPRAYBTLM
Add to Cart
Add to Cart

Multi-use spray bottle holder holds spray bottles and wipes.

Durable and cleanable PETG construction

Specifications

Color Clear
Material PETG
Mounting Type Powerful Magnet
For Use With (Application) Spray bottle
Quantity 1
Height (Metric) 127 mm
Length (Metric) 108 mm
Width (Metric) 203 mm

consumables not included

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.