Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Spray Bottle, Wipe Cannister, Paper Towel Combo Unit
SDP

Catalog No. 22022534
Change view
Click to view available options
Mounting Type:
Magnet mount
Tape mount
Wall mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-022-534 Magnet mount
22-022-533 Tape mount
22-022-532 Wall mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-022-534 Supplier Poltex Supplier No. SPRYPAPRWIPM
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Combo unit holds spray bottles, wipe cannister, and trifold paper towels. Fits up to 4.25 in. diameter wipe cannister. Paper towel stack inserts from the side and dispenses from the front, or underneath.

  • Holds spray bottles, wipe cannister and trifold paper towels
  • Fits up to 4.25in diameter wipe cannister
  • Paper towel stack inserts from the side and dispenses from the front, or underneath
  • Durable and cleanable PETG construction
  • Available as magnet mount (powerful magnets included), wall mount (screws/anchors included), and tape mount (VHB tape included)

Specifications

Product Type Clinical Lab Organizer
No. of Shelves 1
No. of Doors None
Mounting Type Magnet mount
Color Clear
For Use With (Equipment) Spray bottle, wipe cannister, paper towel
Autoclavable Not Autoclavable
Material PETG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.