Learn More
Description
- Adjustable nozzle settings for switching between a fine mist and a targeted stream
- Enables even and controlled distribution of luminol and other detection reagents
- Supports precise application across different investigation requirements
- Pump-operated design for controlled application
- Adjustable nozzle for flexible spray settings
- Designed for even misting of questioned areas
Specifications
Specifications
| Capacity (Metric) | 453.59 g |
| Color | White |
| Material | Plastic |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Chemical Label | Unlabeled |
| Closure Type | Adjustable nozzle |
| Capacity (English) | 16 oz. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
