Learn More
Glass Expansion Spray Chamber Drain Line Agilent ICP-MS
SDP
Supplier:  Glass Expansion 708031476
Encompass_Preferred
Spray chamber drain tubing assembly (G3280-60555, and alternative to G1833-65411). Compatible with quartz Scott style double-pass spray chambers for the Agilent 7700, 7800, 7850, 7900, 8800, 8900 ICP-MS.Push-fit connection to spray chamber drain and barbed connector to peristaltic pump tubing. Used to drain spray chamber waste.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2607546