Learn More
Thermo Scientific Spray Chamber Drain Line
Catalog No. NC1098480 Shop All Thermo Scientific Products
Encompass
Plastic waste drain tubing removes solvents and solutions from spray chamber component of mass spectrometer or inductively coupled plasma system through inert plastic material preventing contamination.
Catalog No. NC1098480 Supplier Thermo Scientific Supplier No. 1320280
Plastic waste drain tubing removes solvents and solutions from spray chamber component of mass spectrometer or inductively coupled plasma system through inert plastic material preventing contamination.