Learn More
Leica Biosystems SPRAY FIX CYTOLOGY FIXAT 12/CS
Catalog No. NC1386849
Encompass
Spray Fix Cytology Fixative Aerosol Container is a high-quality cytology fixative that combines the desirable features of ethyl alcohol with those of polyethylene glycol. The freezing effect that many aerosols have on cells is eliminated because no fluorocarbon propellants are used.
Catalog No. NC1386849 Supplier Leica Biosystems Supplier No. 3801815
Spray Fix Cytology Fixative Aerosol Container is a high-quality cytology fixative that combines the desirable features of ethyl alcohol with those of polyethylene glycol. The freezing effect that many aerosols have on cells is eliminated because no fluorocarbon propellants are used.