Learn More
MSC Spray Nine Heavy-Duty Cleaner, Degreaser, Disinfectant, EPA Registered, 1 gal
Catalog No. 502884721
Encompass
Spray Nine Heavy-Duty Cleaner, Degreaser & Disinfectant is an EPA-registered and NSF approved formula designed for automotive, industrial, military, and DIY markets. This water-based blend provides superior cleaning and degreasing, capable of lifting tough grease, grime, mold, and discolored stains, while also disinfecting hard non-porous surfaces.
  • Tackles tough tasks with less scrubbing
  • Replaces heavy solvent degreasers
  • Kills germs and helps control mold and mildew when used as directed
  • VOC compliant in all 50 states
  • NSF C-1 rated
  • EPA registered (#6659-3)
Catalog No. 50-288-4721 Supplier MSC Supplier No. 63823652
Spray Nine Heavy-Duty Cleaner, Degreaser & Disinfectant is an EPA-registered and NSF approved formula designed for automotive, industrial, military, and DIY markets. This water-based blend provides superior cleaning and degreasing, capable of lifting tough grease, grime, mold, and discolored stains, while also disinfecting hard non-porous surfaces.
  • Tackles tough tasks with less scrubbing
  • Replaces heavy solvent degreasers
  • Kills germs and helps control mold and mildew when used as directed
  • VOC compliant in all 50 states
  • NSF C-1 rated
  • EPA registered (#6659-3)