Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Spray Nozzles for TLC Sprayer

Catalog No. M1085410001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
6 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1085410001 6 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1085410001 Supplier MilliporeSigma™ Supplier No. 1.08541.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for TLC Sprayer

  • Accessory for TLC Sprayer
TRUSTED_SUSTAINABILITY

Specifications

Quantity 6 Units
Type Spray Nozzles for TLC Sprayer
For Use With (Equipment) TLC Sprayer
Storage Requirements Storage temperature: no restrictions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.