Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ SprayPro Inox™

Catalog No. 19200900 Shop All Vileda Professional Products
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-200-900 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-900 Supplier Vileda Professional™ Supplier No. 151514
  • $1,202.00
    $884.65 / Case of 6

    Save $317.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SprayPro Inox™

  • Stainless steel finish and is lightweight and easy to use
  • Controlled water release-3ml with each spray
  • Color coded stickers with each handle
  • Ergonomic hand grip is easy to use in a figure 8 and S movements
  • Cleans over 807 sqft
  • Water/solution is stored in the handle- no external bottles interfering with mopping
  • Simple spray trigger
  • Universal frame connector works with all Vileda Professional cleaning systems
  • Everything contained within the handle - No buckets or wringers required
  • Ideal for sites with limited storage

Specifications

Dimensions 57 x 4.8 x 1.6 in.
Product Type Spray Mop Handle
Color Silver, Blue
Capacity Cleans over 807 sq. ft.
Quantity 6/Cs.
Includes Handle Only
For Use With (Equipment) Any Vileda Professional Mop Frame
Description Professional, ready-to-use Spray Floor Cleaning System
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.