Learn More
Cole-Parmer Cell Spreaders, Sterile, Polypropylene, Individual Wrap; 100/cs
Catalog No. 501960601 Shop All Cole-Parmer Products
Encompass_Preferred
Spread bacterial cultures evenly. Apply cultures over an entire petri dish or plate with the L-shaped spreader. Reduce the risk of damaging the agar-smooth spreading surface with slight upward return.
Catalog No. 50-196-0601 Supplier Cole-Parmer Supplier No. 0439652
Spread bacterial cultures evenly. Apply cultures over an entire petri dish or plate with the L-shaped spreader. Reduce the risk of damaging the agar-smooth spreading surface with slight upward return.