Learn More
Med Vet International Walton Cast Spreader, 9in, 3 Prong, Serrated Jaws, Spring Action, German Grade, Von Klaus
Catalog No. 502092764
Encompass_Preferred
9' Walton cast spreader is a German grade surgical instrument used for spreading apart and remove hard plaster or fiberglass casting materials. It features 3 prongs, serrated jaws, and is spring action for one-hand operation. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2764 Supplier Med Vet International Supplier No. VK1407523
9' Walton cast spreader is a German grade surgical instrument used for spreading apart and remove hard plaster or fiberglass casting materials. It features 3 prongs, serrated jaws, and is spring action for one-hand operation. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.