Learn More
Research Products International Corp SprialPestle Grinding Beads, 50ml
SDP
Catalog No. 501978490
Encompass
Grinding Beads are ideal for quickly and efficiently homogenizing samples in micro-tubes. When used with SpiralPestle Grinder - beads allow optimum cell lysis of culture cells, small tissue samples, and other soft solids. Adding up to 0.1 ml Grinding Beads to a sample and using the SpiralPestle Grinder enhances disruption results in a 1.5 ml micro-tube.
Optional BioVortexer facilitates faster results and efficiency. Combining SpiralPestle Grinder, Beads and BioVortexer greatly increases rotational speed and mechanical dispersement of samples.
Homogenize and grind samples
Allow optimum cell lysis of tissues and microorganisms
Convenient
Catalog No. 50-197-8490 Supplier Research Products International Corp Supplier No. 299017GB
Grinding Beads are ideal for quickly and efficiently homogenizing samples in micro-tubes. When used with SpiralPestle Grinder - beads allow optimum cell lysis of culture cells, small tissue samples, and other soft solids. Adding up to 0.1 ml Grinding Beads to a sample and using the SpiralPestle Grinder enhances disruption results in a 1.5 ml micro-tube.
Optional BioVortexer facilitates faster results and efficiency. Combining SpiralPestle Grinder, Beads and BioVortexer greatly increases rotational speed and mechanical dispersement of samples.
Homogenize and grind samples
Allow optimum cell lysis of tissues and microorganisms
Convenient