Learn More
Fine Science Tools Spring Scissors, Stainless Steel, Angled to Side, 10 cm
SDP
Catalog No. NC9791568
Encompass
Tip Shape: Angled to Side/ Tips: Sharp-Ball Tip/ Cutting Edge: 8mm/ Tip Diameter: 0.125mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No/ Feature: 1.3mm Ball Diameter
Catalog No. NC9791568 Supplier Fine Science Tools Supplier No. 1503309
Tip Shape: Angled to Side/ Tips: Sharp-Ball Tip/ Cutting Edge: 8mm/ Tip Diameter: 0.125mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No/ Feature: 1.3mm Ball Diameter