Learn More
Quality Lab Accessories SPRING CAPSULE SINKER SS
Catalog No. NC9603067
Encompass
Spring Style Capsule Sinker, 316 SS, .91" (23mm) L � 31" (8mm) W capacity
Catalog No. NC9603067 Supplier Quality Lab Accessories Supplier No. CAPWST23
Spring Style Capsule Sinker, 316 SS, .91" (23mm) L � 31" (8mm) W capacity