Learn More
Julabo USA Inc Each, 1L Erlenmeyer spring clamp for SW22/SW23
SDP
Catalog No. 507534092
Encompass_Preferred
Bath Accessories, Julabo,Spring clamps for Erlenmeyer flasks 1L. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 6 1L spring clamps can install on this tray.
Catalog No. 50-753-4092 Supplier Julabo USA Inc Supplier No. 8970609
Bath Accessories, Julabo,Spring clamps for Erlenmeyer flasks 1L. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 6 1L spring clamps can install on this tray.