Learn More
Julabo USA Inc Each, 100 mL Erlenmeyer spring clamp for SW22/SW23
SDP
Catalog No. 507534088
Encompass_Preferred
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 100 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 16 100mL spring clamps can install on this tray.
Catalog No. 50-753-4088 Supplier Julabo USA Inc Supplier No. 8970604
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 100 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 16 100mL spring clamps can install on this tray.