Learn More
Julabo USA Inc Each, 10 mL Erlenmeyer spring clamp for SW22/SW23
SDP
Catalog No. 507534085
Encompass_Preferred
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 10 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 56 10mL spring clamps can install on this tray.
Catalog No. 50-753-4085 Supplier Julabo USA Inc Supplier No. 8970601
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 10 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 56 10mL spring clamps can install on this tray.