Learn More
Julabo USA Inc Each, 200/250 mL Erlenmeyer spring clamp for SW22/SW23
SDP
Catalog No. 507534089
Encompass_Preferred
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 200/250 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 10 200-250 mL spring clamps can install on this tray.
Catalog No. 50-753-4089 Supplier Julabo USA Inc Supplier No. 8970606
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 200/250 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 10 200-250 mL spring clamps can install on this tray.