Learn More
Julabo USA Inc Each, 300 mL Erlenmeyer spring clamp for SW22/SW23
SDP
Catalog No. 507534090
Encompass_Preferred
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 300 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 7 300mL spring clamps can install on this tray.
Catalog No. 50-753-4090 Supplier Julabo USA Inc Supplier No. 8970607
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 300 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 7 300mL spring clamps can install on this tray.