Learn More
Julabo USA Inc Each, 500 mL Erlenmeyer spring clamp for SW22/SW23
SDP
Catalog No. 507534091
Encompass_Preferred
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 500 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 6 500mL spring clamps can install on this tray.
Catalog No. 50-753-4091 Supplier Julabo USA Inc Supplier No. 8970608
Bath Accessories, Julabo, Spring clamps for Erlenmeyer flasks, 500 mL. Installs on basic tray 8970620 (sold separately) for use with shaking water bath models SW22, SW23. A quantity of 6 500mL spring clamps can install on this tray.