Learn More
Julabo USA Inc Each, spring clamp tray for SW22/SW23 shaking water baths
SDP
Catalog No. 507534093
Encompass_Preferred
Bath accessoreies, Julabo, Basic tray for assembling spring clamps. For use with shaking water bath models SW22, SW23. Mix and match combinations of spring clamps 8970601 through 8970609 install on the basic tray.
Catalog No. 50-753-4093 Supplier Julabo USA Inc Supplier No. 8970620
Bath accessoreies, Julabo, Basic tray for assembling spring clamps. For use with shaking water bath models SW22, SW23. Mix and match combinations of spring clamps 8970601 through 8970609 install on the basic tray.