Learn More
Labrepco Inc SPRING CLIP VERTICAL RACKS FOR
SDP
Catalog No. 509020138
Encompass_Preferred
Rack, Vertical; Labrepco; Spring clip; For chest freezers or liquid nitrogen tanks; For standard 2 in. boxes; Made of non-corrosive stainless steel; With spring lock only; L x H x D: 24.187 x 5.625 x 5.5 in.; Capacity: 11
Catalog No. 50-902-0138 Supplier Labrepco Inc Supplier No. CF112S
Rack, Vertical; Labrepco; Spring clip; For chest freezers or liquid nitrogen tanks; For standard 2 in. boxes; Made of non-corrosive stainless steel; With spring lock only; L x H x D: 24.187 x 5.625 x 5.5 in.; Capacity: 11