Learn More
MSC Value Collection Extension Spring, 1/4 in. OD, 0.8125 lb./in. Rate, 12/Pk.
Catalog No. 5000512156
Encompass
This extension spring is designed for applications requiring consistent tension and durability. With an outer diameter of 1/4 in., it provides reliable performance in mechanical assemblies.
  • Outer diameter of 1/4 in.
  • Spring rate of 0.8125 lb/in.
Catalog No. 50-005-12156 Supplier MSC Supplier No. 53620530
This extension spring is designed for applications requiring consistent tension and durability. With an outer diameter of 1/4 in., it provides reliable performance in mechanical assemblies.
  • Outer diameter of 1/4 in.
  • Spring rate of 0.8125 lb/in.