Learn More
Waters Corp Waters Corporation Spring for Low Volume Insert, 4 mL Vial, 144 per package
Catalog No. 50785889 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Vial inserts are used in applications where there is limited sample volume. This product is a spring designed to keep a vial insert secure within a 4 mL vial and protect it from hitting the interior walls of the sample collection vial. It helps ensure that sample preparation protocols are performed successfully, especially when limited sample volume is available. The spring works with vial inserts to reduce the available volume in the vial to a lower surface area, which aids in creating ideal needle conditions for sample withdrawal and maximizes sample retrieval when used with various autosamplers.
  • Keeps vial insert secure within the vial.
  • Protects the vial insert from hitting the interior walls of the sample collection vial.
  • Designed for sample preparation applications with limited sample volume.
Catalog No. 50-785-889 Supplier Waters Corp Supplier No. WAT072708
Vial inserts are used in applications where there is limited sample volume. This product is a spring designed to keep a vial insert secure within a 4 mL vial and protect it from hitting the interior walls of the sample collection vial. It helps ensure that sample preparation protocols are performed successfully, especially when limited sample volume is available. The spring works with vial inserts to reduce the available volume in the vial to a lower surface area, which aids in creating ideal needle conditions for sample withdrawal and maximizes sample retrieval when used with various autosamplers.
  • Keeps vial insert secure within the vial.
  • Protects the vial insert from hitting the interior walls of the sample collection vial.
  • Designed for sample preparation applications with limited sample volume.