Learn More
Med Vet International Spring Infusor, 10mL, J0830, Each
Catalog No. 5000585256
Encompass_Preferred
Springfusor, a simple reusable spring-driver pump used for continuous IV infusions over time, re- quires no batteries or electricity. The pump is matched with various special sized tubing to determine actual flow rate. Uses include adminstering oncology and anesthetic drugs, and IV fluids to very small patients.Features:• Large variety of flow rates available (e.g., 10ml, 15 to 60 minutes)• Compact and very portable• Reusable• No programming required• Not gravity dependentSpring Infusor, 10ml.
Catalog No. 50-005-85256 Supplier Med Vet International Supplier No. JOR830
Springfusor, a simple reusable spring-driver pump used for continuous IV infusions over time, re- quires no batteries or electricity. The pump is matched with various special sized tubing to determine actual flow rate. Uses include adminstering oncology and anesthetic drugs, and IV fluids to very small patients.Features:• Large variety of flow rates available (e.g., 10ml, 15 to 60 minutes)• Compact and very portable• Reusable• No programming required• Not gravity dependentSpring Infusor, 10ml.